Learn More
Ace Glass, Inc. 40mm straight tubes with 25-50 micron fritted discs
SDP
Catalog No. 50974384
Encompass_Preferred
40mm straight tubes with 25-50 micron fritted discs, for construction of special apparatus.
Catalog No. 50-974-384 Supplier Ace Glass, Inc. Supplier No. 720536
40mm straight tubes with 25-50 micron fritted discs, for construction of special apparatus.