Learn More
Ace Glass, Inc. 40mm straight tubes with 10-20 micron fritted discs
SDP
Catalog No. 50979201
Encompass_Preferred
40mm straight tubes with 10-20 micron fritted discs, for construction of special apparatus.
Catalog No. 50-979-201 Supplier Ace Glass, Inc. Supplier No. 720538
40mm straight tubes with 10-20 micron fritted discs, for construction of special apparatus.