Learn More
Azzota Corp 40mm Pathlength Standard Cuvette, 14Ml Pathlength, 40mm Inside Width, 10mm Volume, 14.0Ml Dimensions, 45X12.5X42.5mm, Cover And Round Bottom Quartz (190Nm-2500Nm)
Catalog No. 507532907
Encompass_Preferred
40mm Pathlength Standard Cuvette, 14ml Pathlength, 40mm Inside width, 10mm Volume, 14.0ml Dimensions, 45x12.5x42.5mm, Cover and round bottom Quartz (190nm-2500nm)
Catalog No. 50-753-2907 Supplier Azzota Corp Supplier No. Q107
40mm Pathlength Standard Cuvette, 14ml Pathlength, 40mm Inside width, 10mm Volume, 14.0ml Dimensions, 45x12.5x42.5mm, Cover and round bottom Quartz (190nm-2500nm)