Learn More
Novus Biologicals™ 40nm Colloidal Gold Nanoparticles - 15OD

Description
- Narrow size distribution, consistent shape
- Less prone to aggregation
- Uniform shape to ensures consistent binding of antibodies
Applications: Electron microscopy visualization
Storage: Store at 4°C
Note: Kits are guaranteed for 6 months from date of receipt
Specifications
Specifications
| Quantity | 10 mL |
| Product Type | 40nm Colloidal Gold Nanoparticles - 15OD Antibody Labeling Kit |
| Content And Storage | 4°C Do not freeze. |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.