Learn More
Pluriselect pluriStrainer® 40 µm (Cell Strainer) 25/pk
Catalog No. NC3359348
Encompass
Feature-rich sieving device for filtration and purification of sample material and other biological liquids.The pluriStrainer® is a sieving device (or cell strainer) for a wide range of laboratory applications where the filtration and the purification of liquids is required. Its unique design features improved ventilation to avoid clogging for a smart sample preparation. pluriStrainer® are stackable to allow direct filtration with different mesh sizes and alsocan be inverted to recover the sieved material. For large sample volumes it can be combined with a funnel. It fits all major brand 50 mL centrifuge (conical) tubes. The combination of the pluriStrainer® and the Connector Ring allow using low pressure which supports the filtration. Furthermore it is possible to block the flow through the sieve and hold back the liquid on top of the mesh.
Catalog No. NC3359348 Supplier Pluriselect Supplier No. 435704003
Feature-rich sieving device for filtration and purification of sample material and other biological liquids.The pluriStrainer® is a sieving device (or cell strainer) for a wide range of laboratory applications where the filtration and the purification of liquids is required. Its unique design features improved ventilation to avoid clogging for a smart sample preparation. pluriStrainer® are stackable to allow direct filtration with different mesh sizes and alsocan be inverted to recover the sieved material. For large sample volumes it can be combined with a funnel. It fits all major brand 50 mL centrifuge (conical) tubes. The combination of the pluriStrainer® and the Connector Ring allow using low pressure which supports the filtration. Furthermore it is possible to block the flow through the sieve and hold back the liquid on top of the mesh.