Learn More
Ace Glass, Inc. PTFE banana stir blade, 40x263mm, 22L flask size, 10mmshafts
SDP
Catalog No. 50979668
Encompass_Preferred
40x263mm PTFE banana shaped stir blade for 10mm O.D. button type stir shafts. 3mm thick blades have a number of perforations, and are designed to closely fit a 22L round bottom flask.
Catalog No. 50-979-668 Supplier Ace Glass, Inc. Supplier No. 808719
40x263mm PTFE banana shaped stir blade for 10mm O.D. button type stir shafts. 3mm thick blades have a number of perforations, and are designed to closely fit a 22L round bottom flask.