Learn More
MSC Loctite 233801 1 oz Bottle Clear Instant Adhesive Series 414, 20 sec Fixture Time, 24 hr Full Cure Time, Bonds to Metal, Plastic & Rubber
Supplier:  MSC 88545660
Encompass
Loctite 233801 1 oz Bottle Clear Instant Adhesive Series 414, 20 sec Fixture Time, 24 hr Full Cure Time, Bonds to Metal, Plastic & Rubber
Catalog No. 50-115-0231