Learn More
MSC Loctite 135452 1 oz Bottle Clear Instant Adhesive Series 416, 30 sec Fixture Time, 24 hr Full Cure Time, Bonds to Metal, Plastic & Rubber
Supplier:  MSC 88545728
Encompass
Loctite 135452 1 oz Bottle Clear Instant Adhesive Series 416, 30 sec Fixture Time, 24 hr Full Cure Time, Bonds to Metal, Plastic & Rubber
Catalog No. 50-111-3866