Learn More
Cayman Chemical 41BB LIgnd EXrAClular Do 100ug
SDP
Catalog No. 502949403
Encompass_Preferred
Source: Active recombinant N-terminal human IgG1 Fc-tagged 4-1BB ligand expressed in HEK293 cells • Amino acids: 71-254 • MW: 47.9 kDa
Catalog No. 50-294-9403 Supplier Cayman Chemical Supplier No. 32067100 UG
Source: Active recombinant N-terminal human IgG1 Fc-tagged 4-1BB ligand expressed in HEK293 cells • Amino acids: 71-254 • MW: 47.9 kDa