Learn More
Cayman Chemical 41BBCD137 EXrAClular Dom 100ug
SDP
Catalog No. 502947282
Encompass_Preferred
Source: Recombinant C-terminal human IgG1 Fc-His-tagged 4-1BB expressed in HEK293 cells • Amino acids: 25-186 • MW: 45.2 kDa
Catalog No. 50-294-7282 Supplier Cayman Chemical Supplier No. 31825100 UG
Source: Recombinant C-terminal human IgG1 Fc-His-tagged 4-1BB expressed in HEK293 cells • Amino acids: 25-186 • MW: 45.2 kDa