Learn More
ACROBiosystems Recombinant Protein;Human 4-1BB / TNFRSF9 Protein, Fc Tag;HEK293;1MG;41B,TNFRSF9,4-1BB,CD137,CDw137,ILA,TNFRSF-9,41BB,4-1BBL
Catalog No. 501659081
Encompass_Preferred
Recombinant Protein;Human 4-1BB / TNFRSF9 Protein, Fc Tag;1MG;Human 4-1BB, Fc Tag (41B-H5258) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - Gln 186 (Accession # NP_001552.2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 43.3 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human 4-1BB, Fc Tag (Cat. No. 41B-H5258) at 0.1 μg/mL (100 μL/well) can bind Biotinylated Human 4‑1BB Ligand, Fc Tag, Avi Tag (Cat. No. 41L-H82F9) with a linear range of 0.4-13 ng/mL (QC tested).
Catalog No. 50-165-9081 Supplier ACROBiosystems Supplier No. 41BH52581MG
Recombinant Protein;Human 4-1BB / TNFRSF9 Protein, Fc Tag;1MG;Human 4-1BB, Fc Tag (41B-H5258) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - Gln 186 (Accession # NP_001552.2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 43.3 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human 4-1BB, Fc Tag (Cat. No. 41B-H5258) at 0.1 μg/mL (100 μL/well) can bind Biotinylated Human 4‑1BB Ligand, Fc Tag, Avi Tag (Cat. No. 41L-H82F9) with a linear range of 0.4-13 ng/mL (QC tested).