Learn More
ACROBiosystems Recombinant Protein;Cynomolgus / Rhesus macaque 4-1BB / TNFRSF9 Protein;HEK293;1MG;41B,TNFRSF9,4-1BB,CD137,CDw137,ILA,TNFRSF-9,41BB,4-1BBL
Catalog No. 501659134
Encompass_Preferred
Recombinant Protein;Cynomolgus / Rhesus macaque 4-1BB / TNFRSF9 Protein;1MG;Cynomolgus / Rhesus macaque 4-1BB, His Tag (41B-C52H4) is expressed from human 293 cells (HEK293). It contains AA Leu 24 - Gln 186 (Accession # XP_005544945.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.2 kDa. The protein migrates as 30-40 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Cynomolgus / Rhesus macaque 4-1BB, His Tag (Cat. No. 41B-C52H4) at 1 μg/mL (100 μL/well) can bind Human 4‑1BB Ligand, Fc Tag (Cat. No. 41L-H5257) with a linear range of 0.05-0.5 ng/mL (QC tested).
Catalog No. 50-165-9134 Supplier ACROBiosystems Supplier No. 41BC52H41MG
Recombinant Protein;Cynomolgus / Rhesus macaque 4-1BB / TNFRSF9 Protein;1MG;Cynomolgus / Rhesus macaque 4-1BB, His Tag (41B-C52H4) is expressed from human 293 cells (HEK293). It contains AA Leu 24 - Gln 186 (Accession # XP_005544945.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.2 kDa. The protein migrates as 30-40 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Cynomolgus / Rhesus macaque 4-1BB, His Tag (Cat. No. 41B-C52H4) at 1 μg/mL (100 μL/well) can bind Human 4‑1BB Ligand, Fc Tag (Cat. No. 41L-H5257) with a linear range of 0.05-0.5 ng/mL (QC tested).