Learn More
ACROBiosystems Recombinant Protein;Human 4-1BB / TNFRSF9 Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;1MG;41B,TNFRSF9,4-1BB,CD137,CDw137,ILA,TNFRSF-9,41BB,4-1BBL
Supplier:  ACROBiosystems 41BH52561MG
Encompass_Preferred
Recombinant Protein;Human 4-1BB / TNFRSF9 Protein, Mouse IgG2a Fc Tag, low endotoxin;1MG;Human 4-1BB, Mouse IgG2a Fc Tag, low endotoxin (41B-H5256) is expressed from human 293 cells (HEK293). It contains AA Leu 24 - Gln 186 (Accession # Q07011-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 44.2 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human 4‑1BB Ligand, Fc Tag (Cat. No. 41L-H5257) at 1 μg/mL (100 μL/well) can bind Human 4-1BB, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. 41B-H5256) with a linear range of 0.6-5 ng/mL (QC tested).
Catalog No. 50-165-7998