Learn More
Ace Glass, Inc. 41mm x 51cm chromatography column
SDP
Catalog No. 50978590
Encompass_Preferred
41mm x 51cm chromatography column with 2mm bore 1:5 taper solid PTFE stopcock plug at the bottom.
Catalog No. 50-978-590 Supplier Ace Glass, Inc. Supplier No. 588850
41mm x 51cm chromatography column with 2mm bore 1:5 taper solid PTFE stopcock plug at the bottom.