Learn More
Ace Glass, Inc. 41mm x 61cm chromatography column
SDP
Supplier:  Ace Glass, Inc. 588935
Encompass_Preferred
41mm X 61cm chromatography column with 2mm bore. Also features 1:5 taper solid PTFE stopcock plug and porosity B (70-100m) fritted disc sealed in.
Catalog No. 50-972-556