Learn More
Ace Glass, Inc. 41mm x 61cm chromatography column
SDP
Supplier:  Ace Glass, Inc. 588855
Encompass_Preferred
41mm x 61cm chromatography column with 2mm bore 1:5 taper solid PTFE stopcock plug at the bottom.
Catalog No. 50-972-552