Learn More
Medchemexpress LLC HY-12424A 5mg Medchemexpress, 42-(2-Tetrazolyl)rapamycin CAS:221877-56-1 Purity:>98%
SDP
Catalog No. 502081040
Encompass_Preferred
Medchemexpress, HY-12424A 5mg 42-(2-Tetrazolyl)rapamycin CAS:221877-56-1 42-(2-Tetrazolyl)rapamycin is a prodrug compound of a rapamycin analog extracted from patent US 20080171763 A1, Example 1. Rapamycin is a specific mTOR inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1040 Supplier Medchemexpress LLC Supplier No. HY12424A5MG
Medchemexpress, HY-12424A 5mg 42-(2-Tetrazolyl)rapamycin CAS:221877-56-1 42-(2-Tetrazolyl)rapamycin is a prodrug compound of a rapamycin analog extracted from patent US 20080171763 A1, Example 1. Rapamycin is a specific mTOR inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.