Learn More
Moldex™ 4200 Series AirWave™ N95 Particulate Respirators
SDP
Catalog No. p-4591653 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Size:
Medium/Large
Catalog No. Size
19-999-370 Medium/Large
1 options
Catalog No. 19-999-370 Supplier Moldex™ Supplier No. 4200
Allows more airflow in and out for cool comfort and easier breathing.
  • With unique waves that doubles the filter surface area
  • Dura-Mesh™ shell for bi-directional protection
  • FlexFit™ design expands and contracts while speaking
  • Pinch-free molded nose bridge
  • Softspun™ lining
  • Soft foam nose cushion and contour shaped
  • Available in three different sizes
Specifications
Aerosol Type N - Not Resistant to Oil
ANSI Standard ANSI/ISEA 110-2009, Section 7.11.1
Certifications (NIOSH) NIOSH 42 CFR 84, N95
Design Type Cup/Molded
Disposable Single-use
Filter Type N95
Material Dura-Mesh
Product Type General Purpose Particulate Respirator
Size Medium/Large
With Exhalation Valve No
Filter Efficiency 95%
For Use With (Application) Welding, soldering and brazing, grinding, sanding, sweeping, bagging, stone quarrying, metal machining, cement, underground mining, polishing, buffing, lime, poultry and textiles
Material (Liner) Softspun™
Model 4200N95
Nosepiece Type Molded nose bridge
Quantity 10/Pk.
Strap Attachment Type SmartStrap™
FDA Cleared Surgical Mask No