Learn More
eMolecules​ 42074-68-0 | 2-Chlorotrityl chloride resin (0.4 - 0.8 mmol/g, 100 - 200 mesh) | Chem-Impex | MFCD00040399 | 313.220 | C19H14Cl2 | 0.000 | Clc1ccccc1C(Cl)(c1ccccc1)c1ccccc1 | 100g | 786484296
Catalog No. 5000489956 Shop All eMolecules Inc. Products
Encompass_Preferred
2-Chlorotrityl chloride resin (0.4 - 0.8 mmol/g, 100 - 200 mesh) | Chem-Impex | 42074-68-0 | MFCD00040399 | 313.220 | C19H14Cl2 | 0.000 | Clc1ccccc1C(Cl)(c1ccccc1)c1ccccc1 | 100g | 786484296
Catalog No. 50-004-89956 Supplier eMolecules​ Supplier No. 786484296
2-Chlorotrityl chloride resin (0.4 - 0.8 mmol/g, 100 - 200 mesh) | Chem-Impex | 42074-68-0 | MFCD00040399 | 313.220 | C19H14Cl2 | 0.000 | Clc1ccccc1C(Cl)(c1ccccc1)c1ccccc1 | 100g | 786484296