Learn More
Biolegend Brilliant Violet 421™ anti-mouse CD86
Supplier:  Biolegend 105032
Encompass_Preferred
Brilliant Violet 421™ anti-mouse CD86 [GL-1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. NC1929966