Learn More
Bio Rad Laboratories Model 422 Electro-Eluter
Catalog No. NC1552481
Encompass
Complete electroelution cell, includes buffer tank, lid with power cables, membrane caps (MW limit 12,000–15,000), accessories.
Catalog No. NC1552481 Supplier Bio Rad Laboratories Supplier No. 1652976
Complete electroelution cell, includes buffer tank, lid with power cables, membrane caps (MW limit 12,000–15,000), accessories.