Learn More
MSC 425 3X60YDS SVR ALUM FOIL TAPE
Catalog No. 501108189
Encompass
This 425 Series aluminum foil tape is 3 in wide and 4.6 mil thick with an acrylic adhesive, designed for applications like chemical stripping, splicing, heat reflecting, and sealing.
  • Thermally conductive to dissipate heat and improve heating/cooling efficiency for temperature-sensitive materials
  • Resistant to chemicals, protecting surfaces during de-paint processes and chemical masking
  • Acrylic adhesive offers long-term durability and clean removal after harsh masking applications
  • Withstands exposure to flame, weathering, moisture, and UV
  • Heat and light reflective, protecting surfaces and enhancing lighting efficiency
  • Dead soft aluminum foil backing conforms to curved and uneven surfaces
  • UL Recognized to UL 746C and Classified to UL 723, Class 'L' for low flammability
Catalog No. 50-110-8189 Supplier MSC Supplier No. 31989759
This 425 Series aluminum foil tape is 3 in wide and 4.6 mil thick with an acrylic adhesive, designed for applications like chemical stripping, splicing, heat reflecting, and sealing.
  • Thermally conductive to dissipate heat and improve heating/cooling efficiency for temperature-sensitive materials
  • Resistant to chemicals, protecting surfaces during de-paint processes and chemical masking
  • Acrylic adhesive offers long-term durability and clean removal after harsh masking applications
  • Withstands exposure to flame, weathering, moisture, and UV
  • Heat and light reflective, protecting surfaces and enhancing lighting efficiency
  • Dead soft aluminum foil backing conforms to curved and uneven surfaces
  • UL Recognized to UL 746C and Classified to UL 723, Class 'L' for low flammability