Learn More
CELLTREAT Scientific Products 4250cm2 ESRB Roller Bottle, Vent Cap, Sterile
SDP
Catalog No. NC1772263
Encompass_Preferred
CELLTREAT Scientific Products 4250cm2 Roller Bottle has larger growth area options that maximize valuable laboratory space and save time by reducing repetitive tasks. Ideal for large-scale cell growth. Each bottle has more than twice the cell growth area for 850cm2 bottle. Offers the equivalent growth area of 5 standard 850cm2 bottles, using 40% less plastic. Have uniform hydrophilic surface ideal for consistent cell attachment. Feature horizontal ribs to bathe cells more evenly than bottles with vertical ribs. Large knurled cap for easy grip. Bottles fit into any roller apparatus tha can accomodate 2 standard 850cm2 bottles.
Catalog No. NC1772263 Supplier CELLTREAT Scientific Products Supplier No. 229377
CELLTREAT Scientific Products 4250cm2 Roller Bottle has larger growth area options that maximize valuable laboratory space and save time by reducing repetitive tasks. Ideal for large-scale cell growth. Each bottle has more than twice the cell growth area for 850cm2 bottle. Offers the equivalent growth area of 5 standard 850cm2 bottles, using 40% less plastic. Have uniform hydrophilic surface ideal for consistent cell attachment. Feature horizontal ribs to bathe cells more evenly than bottles with vertical ribs. Large knurled cap for easy grip. Bottles fit into any roller apparatus tha can accomodate 2 standard 850cm2 bottles.