Learn More
Chemglass Life Sciences Dewar Flask, Cylindrical, Wide Mouth, 4300mL Capacity, 143mm ID X 305mm Inside Depth, 178mm OD X 365mm Height
SDP
Catalog No. CG159305
Encompass_Preferred
Dewar Flask, Cylindrical, Wide Mouth, 4300mL Capacity, 143mm ID X 305mm Inside Depth, 178mm OD X 365mm Height. Dewar flask with weighted metal base for stability and protective plastic mesh cover providing a firm grip and a cushion in the event the flask is bumped. Supplied complete with a vented polyethylene stopper to minimize evaporation.
Catalog No. CG159305 Supplier Chemglass Life Sciences Supplier No. CG159305
Dewar Flask, Cylindrical, Wide Mouth, 4300mL Capacity, 143mm ID X 305mm Inside Depth, 178mm OD X 365mm Height. Dewar flask with weighted metal base for stability and protective plastic mesh cover providing a firm grip and a cushion in the event the flask is bumped. Supplied complete with a vented polyethylene stopper to minimize evaporation.