Learn More
eMolecules​ 431041-83-7 | RINK AMIDE MBHA RESIN (100-200 MESH, 1% DVB, 0.3-0.8MMOL/G) | AstaTech732.877 | C47H44N2O6 | 95.000 | COc1ccc(C(NC(=O)OCC2c3ccccc3-c3ccccc23)c2ccc(OCC(=O)NC(c3ccc(C)cc3)c3ccc(C)cc3)cc2)c(OC)c1 | 25g | 784435626
Catalog No. 5000470983 Shop All eMolecules Inc. Products
Encompass_Preferred
RINK AMIDE MBHA RESIN (100-200 MESH, 1% DVB, 0.3-0.8MMOL/G) | AstaTech | 431041-83-7732.877 | C47H44N2O6 | 95.000 | COc1ccc(C(NC(=O)OCC2c3ccccc3-c3ccccc23)c2ccc(OCC(=O)NC(c3ccc(C)cc3)c3ccc(C)cc3)cc2)c(OC)c1 | 25g | 784435626
Catalog No. 50-004-70983 Supplier eMolecules​ Supplier No. 784435626
RINK AMIDE MBHA RESIN (100-200 MESH, 1% DVB, 0.3-0.8MMOL/G) | AstaTech | 431041-83-7732.877 | C47H44N2O6 | 95.000 | COc1ccc(C(NC(=O)OCC2c3ccccc3-c3ccccc23)c2ccc(OCC(=O)NC(c3ccc(C)cc3)c3ccc(C)cc3)cc2)c(OC)c1 | 25g | 784435626