Learn More
Uline Flammable Storage Cabinet (Yellow)
Catalog No. NC1251720
Encompass
45 gallon flammable liquid storage cabinet made of 18 gauge steel with powder coated finish, manual closing doors, three point locking handle, three adjustable shelves, 2 inch leakproof door sill, meets OSHA and fire code requirements for safe flammable liquid storage with fire resistance and ventilation.
Catalog No. NC1251720 Supplier Uline Supplier No. H1564MY
45 gallon flammable liquid storage cabinet made of 18 gauge steel with powder coated finish, manual closing doors, three point locking handle, three adjustable shelves, 2 inch leakproof door sill, meets OSHA and fire code requirements for safe flammable liquid storage with fire resistance and ventilation.