Learn More
Ludlum Measurements Inc 44-9 PANCAKE G-M DETECTOR
SDP
Catalog No. NC9513995
Encompass
Model 44-9 is a pancake detector,INDICATED USE:Alpha beta gamma survey; Frisking,DETECTOR:Pancake type halogen quenched G-M,WINDOW:1.7 plus or minus 0.3 mg/cm2 mica,WINDOW AREA:Active - 15 cm2;Open - 12 cm2
Catalog No. NC9513995 Supplier Ludlum Measurements Inc Supplier No. 471539
Model 44-9 is a pancake detector,INDICATED USE:Alpha beta gamma survey; Frisking,DETECTOR:Pancake type halogen quenched G-M,WINDOW:1.7 plus or minus 0.3 mg/cm2 mica,WINDOW AREA:Active - 15 cm2;Open - 12 cm2