Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cenmed™ 44 Gallon Infectious Waste Bag Liners, Red, 38 x 45 in.
SDP

Catalog No. 21520178
Change view
Click to view available options
Dimensions (L x W):
38 x 45 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
21-520-178 38 x 45 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-520-178 Supplier Cenmed™ Supplier No. 1833600169
Add to Cart
Add to Cart

Infectious waste bag liners constructed of polyethylene.

  • Infectious Waste Liners
  • Biohazard Bag
  • Durable with a 2.5 Mil Thickness
  • Color Red with Black Print
  • Polyethylene
  • Star Seal Closure
  • 38 x 45 in.
  • 44 Gallon Capacity
  • Made in the USA
  • Case of 1000

Specifications

Autoclavable Not Autoclavable
Closure Type Twist Tie
Color Red
Material Polyethylene
Sterility Non-sterile
Dimensions (L x W) 38 x 45 in.
Capacity (English) 40 gal.
Capacity (Metric) 151 L
Length (Metric) 96 cm
Width (Metric) 114 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.