Learn More
Hach Company Natural Water Standard Solution, 3000 ppm Total Dissolved Solids (TDS), 500 mL
Catalog No. 501996993 Shop All Hach Company Products
Encompass
Natural Water Standard Solution. 40% sodium sulfate/40% sodium bicarbonate/20% sodium chloride. TDS: 3000 ppm ±30 ppm, Conductivity: 3960 µS/cm ±39.6 µS/cm at 25°C. 500 mL bottle.
Catalog No. 50-199-6993 Supplier Hach Company Supplier No. 2974849
Natural Water Standard Solution. 40% sodium sulfate/40% sodium bicarbonate/20% sodium chloride. TDS: 3000 ppm ±30 ppm, Conductivity: 3960 µS/cm ±39.6 µS/cm at 25°C. 500 mL bottle.