Learn More
Myron L Company 445 US SOLUTION FOR NAT WATER
SDP
Catalog No. NC0349955
Encompass_Preferred
TDS Calibration Solution 300 PPM, 445 microsiemens gallon size. If a COA is required for this item please also order Fisher part # NC9672510, which is the $40 COA fee. If the COA is requested after the order is placed the fee will increase to $70
Catalog No. NC0349955 Supplier Myron L Company Supplier No. 442300G
TDS Calibration Solution 300 PPM, 445 microsiemens gallon size. If a COA is required for this item please also order Fisher part # NC9672510, which is the $40 COA fee. If the COA is requested after the order is placed the fee will increase to $70