Learn More
MSC 4462 1/2 X72YD FOAM WH 3M DOUB
Catalog No. 501298638
Encompass
3M Double Coated Polyethylene Foam Tapes 4462W combine a conformable closed-cell foam with a rubber adhesive, providing high initial adhesion to various surfaces, including polyethylene and polypropylene. These tapes are ideal for many indoor general purpose mounting and joining applications where moderate temperature and shear performance are required.
  • Foam construction is ideal for joining, mounting, gasketing, and sealing irregular surfaces
  • Bonds effectively to polyethylene, polypropylene, and many other plastics
  • High ultimate adhesion to a wide variety of surfaces
  • Conformable tape bonds to irregular surfaces
  • Unique rubber adhesive system provides quick stick
  • Adheres well to most low surface energy materials
Catalog No. 50-129-8638 Supplier MSC Supplier No. 00188920
3M Double Coated Polyethylene Foam Tapes 4462W combine a conformable closed-cell foam with a rubber adhesive, providing high initial adhesion to various surfaces, including polyethylene and polypropylene. These tapes are ideal for many indoor general purpose mounting and joining applications where moderate temperature and shear performance are required.
  • Foam construction is ideal for joining, mounting, gasketing, and sealing irregular surfaces
  • Bonds effectively to polyethylene, polypropylene, and many other plastics
  • High ultimate adhesion to a wide variety of surfaces
  • Conformable tape bonds to irregular surfaces
  • Unique rubber adhesive system provides quick stick
  • Adheres well to most low surface energy materials