Learn More
MSC 4466 1 X36YD FOAM WH 3M DOUBLE
Catalog No. 501149209
Encompass
3M Double Coated Polyethylene Foam Tape 4466W is a versatile tape featuring a conformable, closed-cell foam backing and a rubber adhesive, engineered for high initial adhesion to a variety of surfaces. This tape is ideal for numerous indoor mounting, joining, gasketing, and sealing applications, especially on irregular surfaces and plastics like polyethylene and polypropylene, where moderate temperature and shear performance are required.
  • Closed cell polyethylene foam carrier
  • Rubber adhesive system
  • Nominal thickness of 62 mil
  • High initial adhesion to various surfaces
  • Conformable to irregular surfaces
  • Quick stick properties
  • Adheres well to low surface energy materials
  • White color for versatile applications
  • Suitable for general purpose indoor mounting and bonding
Catalog No. 50-114-9209 Supplier MSC Supplier No. 00189001
3M Double Coated Polyethylene Foam Tape 4466W is a versatile tape featuring a conformable, closed-cell foam backing and a rubber adhesive, engineered for high initial adhesion to a variety of surfaces. This tape is ideal for numerous indoor mounting, joining, gasketing, and sealing applications, especially on irregular surfaces and plastics like polyethylene and polypropylene, where moderate temperature and shear performance are required.
  • Closed cell polyethylene foam carrier
  • Rubber adhesive system
  • Nominal thickness of 62 mil
  • High initial adhesion to various surfaces
  • Conformable to irregular surfaces
  • Quick stick properties
  • Adheres well to low surface energy materials
  • White color for versatile applications
  • Suitable for general purpose indoor mounting and bonding