Learn More
Ace Glass, Inc. 45/50 10mm debris trap, glass only
SDP
Catalog No. 50975974
Encompass_Preferred
45/50 10mm glass portion for PTFE Trubore debris trap. Use with ACE pstainless steel or polished glass 10mm O.D. stir shafts.
Catalog No. 50-975-974 Supplier Ace Glass, Inc. Supplier No. 8066232
45/50 10mm glass portion for PTFE Trubore debris trap. Use with ACE pstainless steel or polished glass 10mm O.D. stir shafts.