Learn More
Ace Glass, Inc. 45/50 10mm straight adapter, glass only
SDP
Catalog No. 50975959
Encompass_Preferred
45/50 10mm straight adapter, glass portion only, for use with self-aligning, lubricant free PTFE Trubore bearing or combination bearing / debris trap. Use with 10mm O.D. stainless steel or polished glass shafts.
Catalog No. 50-975-959 Supplier Ace Glass, Inc. Supplier No. 806632
45/50 10mm straight adapter, glass portion only, for use with self-aligning, lubricant free PTFE Trubore bearing or combination bearing / debris trap. Use with 10mm O.D. stainless steel or polished glass shafts.