Learn More
Ace Glass, Inc. Complete 45/50 10mm bearing, debris trap style
SDP
Catalog No. 50979647
Encompass_Preferred
45/50 10mm self-aligning, lubricant free PTFE Trubore debris-trap style bearing includes all parts needed. Use with ACE 10mm O.D. stainless steel or polished glass shafts. Bearing is multi-directional, and achieves vacuum down to 1mm Hg.
Catalog No. 50-979-647 Supplier Ace Glass, Inc. Supplier No. 8066332
45/50 10mm self-aligning, lubricant free PTFE Trubore debris-trap style bearing includes all parts needed. Use with ACE 10mm O.D. stainless steel or polished glass shafts. Bearing is multi-directional, and achieves vacuum down to 1mm Hg.