Learn More
Ace Glass, Inc. Connecting adapter, enlarging, 45/50 outer top, 24/40 inner bottom
SDP
Catalog No. 50978272
Encompass_Preferred
Connecting, enlarging adapter featuring a 45/50 outer top female joint and a 24/40 inner bottom male joint.
Catalog No. 50-978-272 Supplier Ace Glass, Inc. Supplier No. 500536
Connecting, enlarging adapter featuring a 45/50 outer top female joint and a 24/40 inner bottom male joint.