Learn More
Ace Glass, Inc. Connecting adapter, enlarging, 45/50 outer top, 29/42 inner bottom
SDP
Catalog No. 50971598
Encompass_Preferred
Connecting, enlarging adapter featuring a 45/50 outer top female joint and a 29/42 inner bottom male joint.
Catalog No. 50-971-598 Supplier Ace Glass, Inc. Supplier No. 500538
Connecting, enlarging adapter featuring a 45/50 outer top female joint and a 29/42 inner bottom male joint.