Learn More
Ace Glass, Inc. PTFE adapter bellows, 45/50 outer top, 45/50 inner bottom155mm length
SDP
Catalog No. 50977986
Encompass
155mm PTFE bellows adapter with 45/50 outer top and inner bottom joints. Bellows is used for correct alignment of standard taper joints and relieves stress in reaction systems. Bellows operates to 200 degrees C.
Catalog No. 50-977-986 Supplier Ace Glass, Inc. Supplier No. 1344136
155mm PTFE bellows adapter with 45/50 outer top and inner bottom joints. Bellows is used for correct alignment of standard taper joints and relieves stress in reaction systems. Bellows operates to 200 degrees C.