Learn More
Ace Glass, Inc. 45/50 500ML MOISTURE RCVR 45/50 500ML MOISTURE RCVR
SDP
Catalog No. 50975350
Encompass_Preferred
500mL moisture receiver with 45/50 joint. Includes a 120¿, 1:5 PTFE stopcock which provides adequate shut-off in the intermediate position. Thermometer joint is 10/30.
Catalog No. 50-975-350 Supplier Ace Glass, Inc. Supplier No. 774719
500mL moisture receiver with 45/50 joint. Includes a 120¿, 1:5 PTFE stopcock which provides adequate shut-off in the intermediate position. Thermometer joint is 10/30.