Learn More
Ace Glass, Inc. 45/50 FLASK HEAD TC 45/50 FLASK HEAD TC
SDP
Catalog No. 50976801
Encompass_Preferred
Standard interchangeable 45/50 Mini-Lab flask head. Head has a 45/50 standard taper polished top joint, and 14/20 standard taper polished side joints. Side arms are placed at the proper angle so that the thermometer or addition tube can be placed in the flask without interfering with the stirring unit.
Catalog No. 50-976-801 Supplier Ace Glass, Inc. Supplier No. 944320
Standard interchangeable 45/50 Mini-Lab flask head. Head has a 45/50 standard taper polished top joint, and 14/20 standard taper polished side joints. Side arms are placed at the proper angle so that the thermometer or addition tube can be placed in the flask without interfering with the stirring unit.