Learn More
Ace Glass, Inc. 45/50 FLASK HEAD 45/50 FLASK HEAD
SDP
Catalog No. 50976800
Encompass_Preferred
Standard interchangeable 45/50 Mini-Lab flask head. Head has a 45/50 standard taper ground top joint, and 14/20 standard taper ground side joints. Side arms are placed at the proper angle so that the thermometer or addition tube can be placed in the flask without interfering with the stirring unit.
Catalog No. 50-976-800 Supplier Ace Glass, Inc. Supplier No. 944310
Standard interchangeable 45/50 Mini-Lab flask head. Head has a 45/50 standard taper ground top joint, and 14/20 standard taper ground side joints. Side arms are placed at the proper angle so that the thermometer or addition tube can be placed in the flask without interfering with the stirring unit.