Learn More
Chemglass Life Sciences Lower Joint Adapter - 45/50 Lower Joint Adapter fits 31mm Stir Shafts, 2-3/8in
SDP
Catalog No. 50871927
Encompass_Preferred
Lower Joint Adapter - 45/50 Lower Joint Adapter fits 31mm Stir Shafts, 2-3/8in. Our NEW mechanical seal bearing is the only design on the market that will permit you to use your current glass, metal or PTFE covered stir shaft. The design allows you to have the flexibility of vertical adjustment and still choose your agitator diameter and style. Bearing has a ferrule and locking nut to ensure that the seal rotates properly, and is supplied complete with a gold plated wrench. Lower joint adapters are supplied complete with perfluorinated o-rings and a loosening nut. PLEASE NOTE: BEARING BODY AND LOWER JOINT ADAPTER MUST BE ORDERED SEPARATELY.**MAX RPM WILL DEPEND ON SHAFT AND/OR AGITATOR TYPE, ALONG WITH ALIGNMENT OF MOTOR AND COUPLING. CHEMGLASS RECOMMENDS A MAX OF 500 RPM WHEN USED WITH ANY GLASS VESSEL, LID OR SHAFT. LONGER LENGTH STIRRER SHAFTS ARE REQUIRED FOR USE WITH CG-2077-M MECHANICAL SEAL STIR BEARINGS.
Catalog No. 50-871-927 Supplier Chemglass Life Sciences Supplier No. CG2077M33
Lower Joint Adapter - 45/50 Lower Joint Adapter fits 31mm Stir Shafts, 2-3/8in. Our NEW mechanical seal bearing is the only design on the market that will permit you to use your current glass, metal or PTFE covered stir shaft. The design allows you to have the flexibility of vertical adjustment and still choose your agitator diameter and style. Bearing has a ferrule and locking nut to ensure that the seal rotates properly, and is supplied complete with a gold plated wrench. Lower joint adapters are supplied complete with perfluorinated o-rings and a loosening nut. PLEASE NOTE: BEARING BODY AND LOWER JOINT ADAPTER MUST BE ORDERED SEPARATELY.**MAX RPM WILL DEPEND ON SHAFT AND/OR AGITATOR TYPE, ALONG WITH ALIGNMENT OF MOTOR AND COUPLING. CHEMGLASS RECOMMENDS A MAX OF 500 RPM WHEN USED WITH ANY GLASS VESSEL, LID OR SHAFT. LONGER LENGTH STIRRER SHAFTS ARE REQUIRED FOR USE WITH CG-2077-M MECHANICAL SEAL STIR BEARINGS.