Learn More
Ace Glass, Inc. Liquid/liquid extractor body, 1300mL, 45/50 top, includes (2)#7 Ace-Thred™; bushings
SDP
Catalog No. 50973452
Encompass
1300mL liquid-liquid extractor body for use with lighter than water extracting solvents. Overflow and boil-up tube uses Ace-Thred connections allowing easy removal for cleaning, reducing cross-contamination among samples. Top joint is 45/50. Extractor includes 2, #7 Ace-Thred bushings.
Catalog No. 50-973-452 Supplier Ace Glass, Inc. Supplier No. 684608
1300mL liquid-liquid extractor body for use with lighter than water extracting solvents. Overflow and boil-up tube uses Ace-Thred connections allowing easy removal for cleaning, reducing cross-contamination among samples. Top joint is 45/50. Extractor includes 2, #7 Ace-Thred bushings.