Learn More
Chemglass Life Sciences 45/50 Outer Joint, 48mm X 2.3W Tubing
SDP
Catalog No. 501531894
Encompass_Preferred
45/50 Outer Joint, 48mm X 2.3W Tubing. Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint.
Catalog No. 50-153-1894 Supplier Chemglass Life Sciences Supplier No. CGB10110
45/50 Outer Joint, 48mm X 2.3W Tubing. Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint.