Learn More
Chemglass Life Sciences 45/50 Outer Joint, Full Length, on 48mm OD x 2.3mm W tubing
SDP
Catalog No. CG10110
Encompass_Preferred
Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint. 45/50 Outer Joint, Full Length, on 48mm OD x 2.3mm W tubing.
Catalog No. CG10110 Supplier Chemglass Life Sciences Supplier No. CG10110
Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint. 45/50 Outer Joint, Full Length, on 48mm OD x 2.3mm W tubing.