Learn More
Ace Glass, Inc. 45/50 PTFE SLEEVE 3/PK 45/50 PTFE SLEEVE 3/PK
SDP
Catalog No. 509557923
Encompass
PTFE sleeves with 45/50 joint size and .4mm wall thickness. Sturdy and reusable, features reinforced gripping ring for easy and safe removal from standard taper ground joints. Pack of 3.
Catalog No. 50-955-7923 Supplier Ace Glass, Inc. Supplier No. 764116
PTFE sleeves with 45/50 joint size and .4mm wall thickness. Sturdy and reusable, features reinforced gripping ring for easy and safe removal from standard taper ground joints. Pack of 3.