Learn More
Ace Glass, Inc. 45/50 PTFE Stopper with Polypropylene Extraction Nut, Solid
SDP
Catalog No. 50977611
Encompass
45/50 standard taper PTFE stopper with polypropylene extraction nut for easy removal from outer joints.
Catalog No. 50-977-611 Supplier Ace Glass, Inc. Supplier No. 1263624
45/50 standard taper PTFE stopper with polypropylene extraction nut for easy removal from outer joints.