Learn More
Chemglass Life Sciences 45/50 PTFE Stopper, 3/4" NPT, Complete with Extraction Nut
SDP
Catalog No. 501531454
Encompass_Preferred
45/50 PTFE Stopper, 3/4" NPT, Complete with Extraction Nut. PTFE stopper adapter is similar to CG-3002-L but with the addition of one female NPT port in the center. The optional PTFE compression fittings provide a convenient way to insert probes or connect PFA tubing or FEP tubing to reactors or flasks. PTFE stopper has an extraction nut permitting easy removal of the stopper from the glass outer joint.
Catalog No. 50-153-1454 Supplier Chemglass Life Sciences Supplier No. CG3002K48
45/50 PTFE Stopper, 3/4" NPT, Complete with Extraction Nut. PTFE stopper adapter is similar to CG-3002-L but with the addition of one female NPT port in the center. The optional PTFE compression fittings provide a convenient way to insert probes or connect PFA tubing or FEP tubing to reactors or flasks. PTFE stopper has an extraction nut permitting easy removal of the stopper from the glass outer joint.