Learn More
Chemglass Life Sciences 45/50 Glass-Filled PTFE Thermocouple Stabilizer with 15 mm Compression Fitting for 1/2 in Pipe Probe (for 150 L Reactors)
SDP
Catalog No. 502733541
Encompass_Preferred
Glass-filled PTFE thermocouple stabilizer designed to support and stabilize 1/2 in pipe thermocouple probes used in larger laboratory reactors. It includes a compression fitting, a removable extension tube that extends below the joint to hold the probe steady, and a side hose barb for inert gas or vacuum connection. Intended for use with 1/2 in pipe coated PT100 sensors in 150 L reactors.
  • Glass-filled PTFE construction for chemical resistance and durability.
  • Compatible with 1/2 in pipe coated PT100 probes.
  • Includes 15 mm compression fitting.
  • Removable extension tube stabilizes the probe below the joint.
  • Side hose barb for inert gas or vacuum connection.
  • Designed for 150 L reactor applications.
Catalog No. 50-273-3541 Supplier Chemglass Life Sciences Supplier No. CG197188
Glass-filled PTFE thermocouple stabilizer designed to support and stabilize 1/2 in pipe thermocouple probes used in larger laboratory reactors. It includes a compression fitting, a removable extension tube that extends below the joint to hold the probe steady, and a side hose barb for inert gas or vacuum connection. Intended for use with 1/2 in pipe coated PT100 sensors in 150 L reactors.
  • Glass-filled PTFE construction for chemical resistance and durability.
  • Compatible with 1/2 in pipe coated PT100 probes.
  • Includes 15 mm compression fitting.
  • Removable extension tube stabilizes the probe below the joint.
  • Side hose barb for inert gas or vacuum connection.
  • Designed for 150 L reactor applications.