Learn More
Chemglass Life Sciences 45/50 TFE Thermocouple Stabilizer, Complete with 14mm Compression Fitting, for use with 1/2" Thermocouple Probe for 75 & 100L Reactors
SDP
Catalog No. 50872396
Encompass_Preferred
45/50 TFE Thermocouple Stabilizer, Complete with 14mm Compression Fitting, for use with 1/2" Thermocouple Probe for 75 & 100L Reactors. Thermocouple Stabilizer for use with 1/2" diameter coated PT100 sensors. Glass filled PTFE 45/50 inner adapter has a 14mm compression fitting and an extension tube that extends below the joint to stabilize the probe when it is inserted into Chemglass 75 & 100L Reactors. The lower extension tube is removable. The side of the adapter is fitted with a hose barb for connection to an inert gas or vacuum.
Catalog No. 50-872-396 Supplier Chemglass Life Sciences Supplier No. CG197187
45/50 TFE Thermocouple Stabilizer, Complete with 14mm Compression Fitting, for use with 1/2" Thermocouple Probe for 75 & 100L Reactors. Thermocouple Stabilizer for use with 1/2" diameter coated PT100 sensors. Glass filled PTFE 45/50 inner adapter has a 14mm compression fitting and an extension tube that extends below the joint to stabilize the probe when it is inserted into Chemglass 75 & 100L Reactors. The lower extension tube is removable. The side of the adapter is fitted with a hose barb for connection to an inert gas or vacuum.